Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU004761

Sigma-Aldrich

MISSION® esiRNA

targeting human CYP27B1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGAAATTCTCGTGTCCCAGACAAAGACATTCATGTGGGTGACTATATTATCCCCAAAAATACGCTGGTCACTCTGTGTCACTATGCCACTTCAAGGGACCCTGCCCAGTTCCCAGAGCCAAATTCTTTTCGTCCAGCTCGCTGGCTGGGGGAGGGTCCCACCCCCCACCCATTTGCATCTCTTCCCTTTGGCTTTGGCAAGCGCAGCTGTATGGGGAGACGCCTGGCAGAGCTTGAATTGCAAATGGCTTTGGCCCAGATCCTAACACATTTTGAGGTGCAGCCTGAGCCAGGTGCGGCCCCAGTTAGACCCAAGACCCGGACTGTCCTGGTACCTGAAAGGAGCATCAACCTACAGTTTTTGGACAGATAGTCCCATGGAAAGAGACTGTCATCATCACCCTTTCATTCATCATAGGGATAAGATTTTTTGTAGGCACAAGACCAAGGTATACATCTTCCCCTAATGCCTATCTGACCAAACTGGATAGAACCACCATAGTGAAGTGTGAGGCGGCCCTGACCAATGTGTGAAGTATGCACTTGGCCTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hana Sustova et al.
Acta physiologica (Oxford, England), 226(3), e13269-e13269 (2019-03-06)
Loss of skeletal muscle is one of the main features of cancer cachexia. Vitamin D (VD) deficiency is associated with impairment of muscle mass and performance and is highly prevalent in cachectic patients; therefore, VD supplementation has been proposed to
Shuo Geng et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 26(5), 1145-1153 (2011-05-05)
1,25-Dihydroxyvitamin D(3)[1,25(OH)(2)D(3)] has many noncalcemic actions that rest on inhibition of proliferation and promotion of differentiation in malignant and normal cell types. 1,25(OH)(2)D(3) stimulates osteoblast differentiation of human marrow stromal cells (hMSCs), but little is known about the effects of
Kaining Liu et al.
PloS one, 7(6), e39878-e39878 (2012-07-05)
We previously demonstrated that 25-hydroxyvitamin D(3), the precursor of 1α,25-dihydroxyvitamin D(3), is abundant around periodontal soft tissues. Here we investigate whether 25-hydroxyvitamin D(3) is converted to 1α,25-dihydroxyvitamin D(3) in periodontal soft tissue cells and explore the possibility of an autocrine/paracrine
Ken-ichiro Tanaka et al.
Biochemical and biophysical research communications, 450(1), 482-487 (2014-06-14)
Vitamin D deficiency and advanced glycation end products (AGEs) are suggested to be involved in the pathogenesis of osteoporosis and sarcopenia. However, the effects of vitamin D and AGEs on myogenesis and the interaction between muscle and bone remains still
Lars Brodowski et al.
PloS one, 9(6), e98527-e98527 (2014-06-03)
Placenta-derived circulating factors contribute to the maternal endothelial dysfunction underlying preeclampsia. Endothelial colony forming cells (ECFC), a sub-population of endothelial progenitor cells (EPCs), are thought to be involved in vasculogenesis and endothelial repair. Low vitamin D concentrations are associated with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service