Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU003101

Sigma-Aldrich

MISSION® esiRNA

targeting human EXOC2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CATGACGCAGAAACTGGAGAATGTTCTGAACAGAGCAAGTAATACTGCAGACACATTGTTTCAAGAAGTATTAGGTCGGAAAGACAAGGCAGATTCCACTAGAAATGCACTCAATGTGCTTCAGCGATTTAAGTTTCTTTTCAACCTTCCTCTAAATATTGAAAGGAATATTCAAAAGGGTGATTATGATGTGGTTATTAATGATTATGAAAAGGCCAAGTCACTTTTTGGGAAAACGGAGGTGCAAGTTTTCAAGAAATATTATGCTGAAGTAGAAACAAGGATTGAAGCTTTAAGAGAATTACTTCTGGATAAATTGCTTGAGACACCATCAACTTTACATGACCAAAAACGTTACATAAGGTACCTGTCTGACCTTCATGCGTCTGGTGACCCTGCTTGGCAATGCATTGGAGCCCAACACAAGTGGATCCTTCAGCTCATGCACAGTTGCAAAGAGGGCTACGTGAAAGATCTGAAAGGCAAGGATTTCTCTTCCAATGTTCCCATGTTGGATCTTGATAATGATACACGTCCCTCAGTGTTGGGCCATCTCAGTC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen-Wen Gu et al.
Reproduction (Cambridge, England), 159(1), 59-71 (2019-11-11)
The invasion of maternal decidua by extravillous trophoblast (EVT) is essential for the establishment and maintenance of pregnancy, and abnormal trophoblast invasion could lead to placenta-associated pathologies including early pregnancy loss and preeclampsia. SEC5, a component of the exocyst complex
Long Yang et al.
BMC biology, 16(1), 46-46 (2018-04-29)
Candida albicans (C. albicans) invasion triggers antifungal innate immunity, and the elevation of cytoplasmic Ca2+ levels via the inositol 1,4,5-trisphosphate receptor (InsP3R) plays a critical role in this process. However, the molecular pathways linking the InsP3R-mediated increase in Ca2+ and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service