Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU002541

Sigma-Aldrich

MISSION® esiRNA

targeting human CXCL10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCACGTGTTGAGATCATTGCTACAATGAAAAAGAAGGGTGAGAAGAGATGTCTGAATCCAGAATCGAAGGCCATCAAGAATTTACTGAAAGCAGTTAGCAAGGAAAGGTCTAAAAGATCTCCTTAAAACCAGAGGGGAGCAAAATCGATGCAGTGCTTCCAAGGATGGACCACACAGAGGCTGCCTCTCCCATCACTTCCCTACATGGAGTATATGTCAAGCCATAATTGTTCTTAGTTTGCAGTTACACTAAAAGGTGACCAATGATGGTCACCAAATCAGCTGCTACTACTCCTGTAGGAAGGTTAATGTTCATCATCCTAAGCTATTCAGTAATAACTCTACCCTGGCACTATAATGTAAGCTCTACTGAGGTGCTATGTTCTTAGTGGATGTTCTGACCCTGCTTCAAATATTTCCCTCACCTTTCCCATCTTCCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Fu et al.
International immunopharmacology, 75, 105783-105783 (2019-08-04)
Myrothecine A, characterized from the extracts of myrothecium roridum strain IFB-E012, isolated as endophytic fungi found in the traditional Chinese medicinal plant Artemisia annua. Here we investigated its roles on anti-tumor and immune regulation in vitro. Dendritic cells (DCs) are
Katrina K Au et al.
Gynecologic oncology, 145(3), 436-445 (2017-03-21)
We recently established that high STAT1 expression and associated T helper type I tumour immune microenvironment (TME) are prognostic and chemotherapy response predictive biomarkers in high-grade serous ovarian cancer (HGSC). STAT1 induced chemokine CXCL10 is key to the recruitment of
Xiuming Wu et al.
Molecular and cellular endocrinology, 512, 110866-110866 (2020-05-18)
Although 70% of estrogen receptor (ER)-positive breast cancer patients can benefit from tamoxifen therapy, the rapid development of tamoxifen resistance hampers the treatment advantage. In this investigation, we found that the serum level of CXCL10 in breast cancer patients was
Jon Cogan et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 22(10), 1741-1752 (2014-08-27)
Patients with recessive dystrophic epidermolysis bullosa (RDEB) have severe, incurable skin fragility, blistering, and multiple skin wounds due to mutations in the gene encoding type VII collagen (C7), the major component of anchoring fibrils mediating epidermal-dermal adherence. Nearly 10-25% of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service