Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E8761

Sigma-Aldrich

Eosin B

Dye content ≥85 %, certified by the Biological Stain Commission

Synonym(s):

4′,5′-Dibromo-2′,7′-dinitrofluorescein disodium salt, Acid Red 91, Eosin Scarlet, Eosin bluish, Saffrosine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C20H6N2O9Br2Na2
CAS Number:
Molecular Weight:
624.06
Colour Index Number:
45400
Beilstein:
4121964
EC Number:
MDL number:
UNSPSC Code:
12171500

grade

certified by the Biological Stain Commission

composition

Dye content, ≥85%

SMILES string

[Na+].[Na+].[O-]C(=O)c1ccccc1C2=C3C=C(C(=O)C(Br)=C3Oc4c(Br)c([O-])c(cc24)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C20H8Br2N2O9.2Na/c21-14-16(25)11(23(29)30)5-9-13(7-3-1-2-4-8(7)20(27)28)10-6-12(24(31)32)17(26)15(22)19(10)33-18(9)14;;/h1-6,25H,(H,27,28);;/q;2*+1/p-2

InChI key

GYYTYUGGVBYJHE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Suitability

Certified for use as a counterstain for alum-hematoxylin and with Lillie′s modified Nocht technique.

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis and testing of biological stains--the Biological Stain Commission Procedures.
D P Penney et al.
Biotechnic & histochemistry : official publication of the Biological Stain Commission, 77(5-6), 237-275 (2003-02-05)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service