Skip to Content
MilliporeSigma
All Photos(1)

Documents

E8625

Sigma-Aldrich

Eserine hemisulfate salt

Synonym(s):

Physostigmine hemisulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21N3O2 · 0.5H2SO4
CAS Number:
Molecular Weight:
324.39
Beilstein:
3828413
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

2-8°C

SMILES string

OS(O)(=O)=O.CNC(=O)Oc1ccc2N(C)[C@H]3N(C)CC[C@@]3(C)c2c1.CNC(=O)Oc4ccc5N(C)[C@H]6N(C)CC[C@@]6(C)c5c4

InChI

1S/2C15H21N3O2.H2O4S/c2*1-15-7-8-17(3)13(15)18(4)12-6-5-10(9-11(12)15)20-14(19)16-2;1-5(2,3)4/h2*5-6,9,13H,7-8H2,1-4H3,(H,16,19);(H2,1,2,3,4)/t2*13-,15+;/m11./s1

InChI key

YYBNDIVPHIWTPK-KYJQVDHRSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Acetylcholinesterase inhibitor that crosses the blood-brain barrier and forms a carbamylated enzyme complex with acetyl cholinesterase that degrades slowly.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y H Lu et al.
Journal of insect science (Online), 13, 9-9 (2013-07-25)
The aphids Sitobion avenae (Fabricius) and Rhopalosiphum padi (Linnaeus) (Hemiptera: Aphidiae) are serious pests on grain crops and usually coexist on late period of wheat growth in China. Bioassays showed that R. padi was more susceptible than S. avenae to
Vikas Trivedi et al.
Biomedical chromatography : BMC, 34(3), e4777-e4777 (2019-12-12)
The article describes a systematic study to overcome the matrix effect during chromatographic analysis of gemfibrozil, rivastigmine, telmisartan and tacrolimus from biological fluids using LC-ESI-MS/MS. All four methods were thoroughly developed by the appropriate choice of analytical column, elution mode
Anderson Ribeiro-Carvalho et al.
PloS one, 15(10), e0239017-e0239017 (2020-10-03)
Organophosphates are among the most used pesticides. Particularly, chlorpyrifos (CPF) is responsible for a number of deleterious effects on brain development, which may program behavioral changes later in life. Here, we investigated whether a regimen of early low level CPF
Jan-Oliver Hollnagel et al.
iScience, 23(7), 101316-101316 (2020-07-13)
Lactate shuttled from blood, astrocytes, and/or oligodendrocytes may serve as the major glucose alternative in brain energy metabolism. However, its effectiveness in fueling neuronal information processing underlying complex cortex functions like perception and memory is unclear. We show that sole
Bruno Nunes et al.
Environmental science and pollution research international, 24(13), 12155-12167 (2017-03-30)
Biomonitoring of chemical contamination requires the use of well-established and validated tools, including biochemical markers that can be potentially affected by exposure to important environmental toxicants. Cholinesterases (ChEs) are present in a large number of species and have been successfully

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service