Skip to Content
MilliporeSigma
All Photos(1)

Documents

E7880

Sigma-Aldrich

Eledoisin

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H85N13O15S
CAS Number:
Molecular Weight:
1188.40
MDL number:
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CCC(C)C(NC(=O)C(Cc1ccccc1)NC(=O)C(C)NC(=O)C(CC(O)=O)NC(=O)C(CCCCN)NC(=O)C(CO)NC(=O)C2CCCN2C(=O)C3CCC(=O)N3)C(=O)NCC(=O)NC(CC(C)C)C(=O)NC(CCSC)C(N)=O

Amino Acid Sequence

pGlu-Pro-Ser-Lys-Asp-Ala-Phe-Ile-Gly-Leu-Met-NH2

Biochem/physiol Actions

Tachykinin originally isolated from the salivary gland of octopus with structural homology to substance P; potent vasodilator and hypotensive agent; induces salivation and increases capillary permeability. Agonist at tachykinin NK-2/NK-3 receptors.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Hölzemann et al.
International journal of peptide and protein research, 44(2), 105-111 (1994-08-01)
The synthesis of 11 cyclic hexapeptides, some of which contain a carbohydrate side chain moiety, is described in this paper. A glycosylamine was coupled without hydroxyl protecting groups either directly or via a butyric acid spacer to the side chain
U Gether et al.
The Journal of biological chemistry, 268(11), 7893-7898 (1993-04-15)
The NK1 (substance P) and NK3 (neurokinin B) receptors are G protein-coupled receptors sharing approximately 70% identity within the membrane-spanning domains. However, they each have a distinct pharmacological profile in respect of peptide binding. To identify epitopes that determine their
M Bergstrom et al.
Scandinavian journal of clinical and laboratory investigation, 55(8), 679-689 (1995-12-01)
Tachykinins are a family of peptides that may be present in and secreted from carcinoid tumours of mid-gut origin. They are likely to play a role in the pathogenesis of, e.g. the flush, dyspnoea and valvular heart disease seen in
P Björup et al.
Bioorganic & medicinal chemistry, 6(7), 891-901 (1998-09-08)
The influence of different reaction systems on alpha-chymotrypsin-catalyzed synthesis of eledoisin and LH-RH peptides from (7 + 4) and (5 + 5) fragments was investigated. The peptide yield was determined in the following systems: buffered aqueous media, frozen solutions, organic
R J Lukas et al.
Neurochemical research, 21(10), 1245-1257 (1996-10-01)
Nicotinic acetylcholine receptors (nAChR) are diverse members of the ligand-gated ion channel superfamily of neurotransmitter receptors and play critical roles in chemical signaling throughout the nervous system. Reports of effects of substance P (SP) on nAChR function prompted us to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service