Skip to Content
MilliporeSigma
All Photos(1)

Documents

E5766

Sigma-Aldrich

(±)−14,15-Epoxyeicosa-(5Z,8Z,11Z)-trienoic acid

~100 μg/mL in ethanol, ≥98%

Synonym(s):

(±)14,15-EpETrE, (±)−14,15-EET

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32O3
Molecular Weight:
320.47
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98%

concentration

~100 μg/mL in ethanol

lipid type

unsaturated FAs

storage temp.

−20°C

SMILES string

CCCCCC1OC1C\C=C/C\C=C/C\C=C/CCCC(O)=O

InChI

1S/C20H32O3/c1-2-3-12-15-18-19(23-18)16-13-10-8-6-4-5-7-9-11-14-17-20(21)22/h4,6-7,9-10,13,18-19H,2-3,5,8,11-12,14-17H2,1H3,(H,21,22)/b6-4-,9-7-,13-10-

InChI key

JBSCUHKPLGKXKH-ILYOTBPNSA-N

Packaging

Packaged under argon

Caution

Unstable to oxygen and direct sunlight, therefore extreme care must be taken in handling this product. All glassware and stainless steel syringe needles to be used in the transfer of the solution should be pre-rinsed with absolute ethanol and air dried. Transfer and removal of samples should be carried out under an inert atmosphere, keeping the bulk at 0°C or below whenever possible.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup - Solvent

Flash Point(C)

14.0 °C - closed cup - Solvent

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service