E4788
Plant RNA/DNA Purification Kit
sufficient for 50 purifications
About This Item
Recommended Products
usage
sufficient for 50 purifications
Quality Level
greener alternative product characteristics
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
greener alternative category
General description
Application
- Real-time PCR
- Southern blotting
- SNP analysis
- Sequencing
- AFLP
- RFLP
- RAPD
- Microsatellite analysis
- qRT-PCR
- Primer extension
- Expression array assays
- Next Generation Sequencing
Features and Benefits
- Total RNA and genomic DNA are column purified simultaneously in under 30 minutes.
- Reduces variability as RNA and DNA are isolated from a single plant sample.
- Isolate nucleic acids from small plant samples.
- Isolates total RNA including microRNAs.
- No phenol/chloroform extraction or alcohol precipitation.
- Isolate DNA-free plant RNA or RNA-free plant DNA
Other Notes
All solutions should be kept tightly sealed and stored at room temperature. These reagents should remain stable for at least 1 year in their unopened containers.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
The Plant/Fungi DNA Isolation Kit provides a rapid method for the isolation and purification of total DNA from a wide range of plant and fungi species.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service