Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

E4505

Sigma-Aldrich

cis-11,14,17-Eicosatrienoic acid sodium salt

~98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C20H33O2Na
CAS Number:
Molecular Weight:
328.46
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

~98%

storage temp.

−20°C

SMILES string

[Na].CC\C=C/C\C=C/C\C=C/CCCCCCCCCC(O)=O

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service