Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E4132

Sigma-Aldrich

1,N6-Etheno-2′-deoxyadenosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13N5O3
CAS Number:
Molecular Weight:
275.26
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
Pricing and availability is not currently available.

form

solid

storage temp.

−20°C

SMILES string

OCC1OC(CC1O)n2cnc3c2ncn4ccnc34

InChI

1S/C12H13N5O3/c18-4-8-7(19)3-9(20-8)17-6-14-10-11-13-1-2-16(11)5-15-12(10)17/h1-2,5-9,18-19H,3-4H2

InChI key

XQQIMTUYVDUWKJ-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mayura Meerang et al.
Free radical biology & medicine, 44(10), 1863-1868 (2008-03-18)
Thalassemic diseases including homozygous beta-thalassemia and beta-thalassemia/Hb E (beta-Thal/Hb E) are prevalent in Southeast Asia. Iron overload is a common complication in beta-thalassemia patients which induces intracellular oxidative stress and lipid peroxidation (LPO). LPO end products generate miscoding etheno adducts
Plamen P Christov et al.
Chemical research in toxicology, 23(8), 1330-1341 (2010-06-29)
Oligonucleotides were synthesized containing the 7-(2-oxoheptyl)-etheno-dGuo adduct, which is derived from the reaction of dGuo and the lipid peroxidation product 4-oxo-2-nonenal. The in vitro replication of 7-(2-oxoheptyl)-etheno-dGuo by the model Y-family polymerase Sulfolobus solfataricus P2 DNA Polymerase IV (Dpo4) was
Jagadeesan Nair et al.
Biochemical and biophysical research communications, 336(2), 478-482 (2005-09-03)
Reactive oxygen species (ROS) and lipid peroxidation (LPO) play a role in aging and degenerative diseases. To correlate oxidative stress and LPO-derived DNA damage, we determined etheno-DNA-adducts in liver and brain from ROS overproducing OXYS rats in comparison with age-matched
Poh-Gek Forkert et al.
Drug metabolism and disposition: the biological fate of chemicals, 35(5), 713-720 (2007-02-14)
Vinyl carbamate (VC) is derived from ethyl carbamate, a carcinogen formed in fermentation of food and alcoholic products. We have undertaken studies to test the hypothesis that an epoxide generated from VC oxidation leads to formation of 1,N6-ethenodeoxyadenosine (epsilon dAS).
Qingming Fang et al.
Mutation research, 624(1-2), 71-79 (2007-05-25)
Intake of linoleic acid (LA) increased etheno-DNA adducts induced by lipid peroxidation (LPO) in white blood cells (WBC) of female but not of male volunteers [J. Nair, C.E. Vaca, I. Velic, M. Mutanen, L.M. Valsta, H. Bartsch, High dietary omega-6

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service