Skip to Content
MilliporeSigma
All Photos(1)

Documents

E2262

Sigma-Aldrich

Estriol 3-carboxymethyl ether

Synonym(s):

1,3,5-(10)-Estratriene-3,16α,17β-triol 3-carboxymethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H26O5
CAS Number:
Molecular Weight:
346.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

2-8°C

SMILES string

[H][C@]12CC[C@]3(C)[C@@H](O)[C@H](O)C[C@@]3([H])[C@]1([H])CCc4cc(OCC(O)=O)ccc24

InChI

1S/C20H26O5/c1-20-7-6-14-13-5-3-12(25-10-18(22)23)8-11(13)2-4-15(14)16(20)9-17(21)19(20)24/h3,5,8,14-17,19,21,24H,2,4,6-7,9-10H2,1H3,(H,22,23)/t14-,15-,16+,17-,19+,20+/m1/s1

InChI key

SZBWYTRVLQFZGU-UQZPWQSVSA-N

Application

Estriol 3-carboxymethyl ether was coupled to BSA and used as immunogen for raising anti-estriol immunoglobulins.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service