Skip to Content
MilliporeSigma

E2256

[des-Tyr1]-Methionine enkephalin

≥97% (HPLC)

Synonym(s):

Gly-Gly-Phe-Met

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H26N4O5S
CAS Number:
Molecular Weight:
410.49
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CSCC[C@H](NC(=O)[C@H](Cc1ccccc1)NC(=O)CNC(=O)CN)C(O)=O

Application

A tetrapeptide important for the study of enkephalinase inhibition

Biochem/physiol Actions

Enkephalinase inhibitor.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Inhibitory potency of various peptides on enkephalinase activity from mouse striatum.
M C Fournie-Zaluski et al.
Biochemical and biophysical research communications, 91(1), 130-135 (1979-11-14)
High-affinity enkephalin-degrading peptidase in brain is increased after morphine.
B Malfroy et al.
Nature, 276(5687), 523-526 (1978-11-30)