Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E137

Sigma-Aldrich

IRL-1620

powder

Synonym(s):

N-Succinyl-[Glu9, Ala11,15]-Endothelin 1 fragment 8-21

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C86H117N17O27
CAS Number:
Molecular Weight:
1820.95
MDL number:
UNSPSC Code:
12352200

form

powder

color

white

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Suc-Asp-Glu-Glu-Ala-Val-Tyr-Phe-Ala-His-Leu-Asp-Ile-Ile-Trp

Biochem/physiol Actions

Selective ETB endothelin receptor agonist.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lívia S M Mesquita et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 148(1), 61-67 (2008-04-26)
Endothelins (ETs) are vasoactive peptides evolutionary well conserved that exert their effects through two specific receptors (ET(A) and ET(B)) widely distributed in all vertebrates. In snakes, the presence and function of endothelins and their receptors are still scarcely described. We
Julie Labonté et al.
Canadian journal of physiology and pharmacology, 86(8), 516-525 (2008-09-02)
We hypothesized that constitutive endothelial NO synthase (eNOS) and inducible NO synthase (iNOS) have opposite effects on the regulation of endothelin and its receptors. We therefore sought to determine whether deletions of iNOS or eNOS genes in mice modulate pressor
Y Fujitani et al.
The Journal of pharmacology and experimental therapeutics, 267(2), 683-689 (1993-11-01)
The signal transduction pathways of endothelin (ET)-induced vasorelaxation in rat aorta were investigated. An agonist for ETB receptors, IRL 1620, induced transient increases in cytosolic Ca++ (peak at about 10 sec) and cyclic GMP (peak at about 20 sec) accompanied
Mary G Leonard et al.
Pharmacological research, 60(5), 402-410 (2009-08-12)
IRL-1620, a highly selective ET(B) receptor agonist, is presently in a phase I clinical trial (NCT00613691) in the United States for patients with recurrent or progressive carcinoma. The effect of acute repeated administration of IRL-1620 on the development of tachyphylaxis
T Watakabe et al.
Biochemical and biophysical research communications, 185(3), 867-873 (1992-06-30)
Suc-[Glu9,Ala11,15]-endothelin(ET)-1(8-21), IRL 1620, is a linear ET-analog specific for the ET-isopeptide-nonselective ETB receptor. The radio-iodinated analog, [125I]IRL 1620, showed a single class of saturable binding to the ETB receptors in porcine lung membranes with a Kd of 18 pM and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service