Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E131

Sigma-Aldrich

Ergocornine

>95%, solid

Synonym(s):

12′-Hydroxy-2′,5′-α-bis(1-methyl-ethyl)ergotaman-3′,6′,18-trione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H39N5O5
CAS Number:
Molecular Weight:
561.67
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

>95%

form

solid

optical activity

[α]/D −105.6°, c = 0.85 in pyridine(lit.)

color

white

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 0.4 mg/mL
ethanol: 1.02 mg/mL
DMSO: 6.5 mg/mL
H2O: insoluble

SMILES string

[H][C@@]12Cc3c[nH]c4cccc(C1=C[C@H](CN2C)C(=O)N[C@@]5(O[C@]6(O)N([C@@H](C(C)C)C(=O)N7CCC[C@@]67[H])C5=O)C(C)C)c34

InChI

1S/C31H39N5O5/c1-16(2)26-28(38)35-11-7-10-24(35)31(40)36(26)29(39)30(41-31,17(3)4)33-27(37)19-12-21-20-8-6-9-22-25(20)18(14-32-22)13-23(21)34(5)15-19/h6,8-9,12,14,16-17,19,23-24,26,32,40H,7,10-11,13,15H2,1-5H3,(H,33,37)/t19-,23-,24+,26+,30-,31+/m1/s1

InChI key

UJYGDMFEEDNVBF-OGGGUQDZSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Dopamine receptor agonist; ergot alkaloid.

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lorenzo Celio et al.
Frontiers in chemistry, 9, 626630-626630 (2021-03-23)
A novel flow injection microfluidic immunoassay system for continuous monitoring of saxitoxin, a lethal biotoxin, in seawater samples is presented in this article. The system consists of a preimmobilized G protein immunoaffinity column connected in line with a lab-on-chip setup.
J T Uilenbroek
Journal of reproduction and fertility, 86(1), 327-333 (1989-05-01)
Injection of an ovulation-blocking dose of pentobarbitone sodium given in the early afternoon of pro-oestrus in rats decreased follicular oestradiol production in vitro the next day (2.42 +/- 0.11 ng/4 h/follicle in pro-oestrous rats, 0.49 +/- 0.04 ng/4 h/follicle in
Madeleine Gross et al.
Journal of AOAC International, 101(3), 618-626 (2017-10-02)
Four sets of polyclonal antibodies against ergot alkaloids ergometrine, ergotamine, α-ergocryptine, and ergocornine were produced and characterized in a competitive direct or indirect enzyme immunoassay (EIA). Standard curve LODs were 0.03 ng/mL (ergometrine EIA) to 2.0 ng/mL (ergocornine EIA). Three
Karl De Ruyck et al.
Environment international, 137, 105539-105539 (2020-02-09)
The European Food Consumption Validation (EFCOVAL) project includes 600 men and women from Belgium, the Czech Republic, France, the Netherlands, and Norway, who had given serum and 24-hour urine samples, and completed 24-hour dietary recall (24-HDR) interviews. Consumption, according to
H Tomogane et al.
Journal of reproduction and fertility, 88(2), 419-425 (1990-03-01)
The length of the first spontaneous oestrous cycle in pubertal Wistar-Imamichi strain rats determined by vaginal smears varied from 5 to 18 days. The variation was ascribed to the period (3-16 days) of the stage of vaginal smears consisting of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service