Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E125

Sigma-Aldrich

β-Estradiol

Synonym(s):

1,3,5-Estratriene-3,17β-diol, 17β-Estradiol, 3,17β-Dihydroxy-1,3,5(10)-estratriene, Dihydrofolliculin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24O2
CAS Number:
Molecular Weight:
272.38
Beilstein:
1914275
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

mp

176-180 °C (lit.)

storage temp.

2-8°C

SMILES string

O[C@H]1CC[C@@]2([H])[C@]3([H])CCC4=CC(O)=CC=C4[C@@]3([H])CC[C@@]21C

InChI

1S/C18H24O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-17,19-20H,2,4,6-9H2,1H3/t14-,15-,16+,17+,18+/m1/s1

InChI key

VOXZDWNPVJITMN-ZBRFXRBCSA-N

Gene Information

human ... ESR1(2099)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

The major estrogen secreted by the premenopausal ovary. Estrogens direct the development of the female phenotype in embryogenesis and during puberty by regulating gene transcription and, thus, protein synthesis. It also induces the production of gonadotropins which, in turn, induce ovulation. Exposure to estradiol increases breast cancer incidence and proliferation.

replaced by

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pascal Pigeon et al.
Journal of medicinal chemistry, 48(8), 2814-2821 (2005-04-15)
A series of ruthenocene derivatives, 1-[4-(O(CH(2))(n)()N(CH(3))(2))phenyl]-1-(4-hydroxyphenyl)-2-ruthenocenylbut-1-ene, with n = 2-5, based on the structure of the breast cancer drug tamoxifen has been prepared. These compounds were obtained, via a McMurry cross-coupling reaction, as a mixture of Z and E isomers
Hirotaka Ishii et al.
The Neuroscientist : a review journal bringing neurobiology, neurology and psychiatry, 13(4), 323-334 (2007-07-24)
It is believed that sex hormones are synthesized in the gonads and reach the brain via the blood circulation. In contrast with this view, the authors have demonstrated that sex hormones are also synthesized locally in the hippocampus and that
Steffen Renner et al.
Nature chemical biology, 5(8), 585-592 (2009-06-30)
The structure- and chemistry-based hierarchical organization of library scaffolds in tree-like arrangements provides a valid, intuitive means to map and navigate chemical space. We demonstrate that scaffold trees built using bioactivity as the key selection criterion for structural simplification during
Mark A Tarnopolsky
Medicine and science in sports and exercise, 40(4), 648-654 (2008-03-05)
Women oxidize more lipid and less carbohydrate and protein compared with men during endurance exercise. The increase in fat oxidation is associated with higher intramyocellular lipid content and use as well as greater adipocyte lipolysis. Glucose rates of appearance and
James S Wright et al.
Journal of medicinal chemistry, 54(2), 433-448 (2010-12-31)
Long-term use of estrogen supplements by women leads to an increased risk of breast and uterine cancers. Possible mechanisms include metabolism of estradiol and compounds related to tumor-initiating quinones, and ligand-induced activation of the estrogen receptors ERα and ERβ which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service