Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E0761

Sigma-Aldrich

Ebelactone A microbial

esterase inhibitor

Synonym(s):

3,11-Dihydroxy-2,4,6,8,10,12-hexamethyl-9-oxo-6-tetradecenoic acid 1,3-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H34O4
CAS Number:
Molecular Weight:
338.48
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

Streptomyces sp.

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

methanol: 10 mg/mL, clear, colorless

storage temp.

2-8°C

SMILES string

[H][C@@]1([C@H](C/C(C)=C/[C@H](C([C@H]([C@@H]([C@@H](CC)C)O)C)=O)C)C)OC([C@H]1C)=O

InChI

1S/C20H34O4/c1-8-12(3)17(21)15(6)18(22)13(4)9-11(2)10-14(5)19-16(7)20(23)24-19/h9,12-17,19,21H,8,10H2,1-7H3/b11-9+/t12-,13-,14+,15+,16+,17-,19+/m1/s1

InChI key

WOISDAHQBUYEAF-QIQXJRRPSA-N

General description

Esterase inhibitors produced by Streptomyces sp. MG7-G1 strain

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amit K Mandal
Organic letters, 4(12), 2043-2045 (2002-06-07)
[structure: see text] The highly stereocontrolled hydroboration of an alkene, a subsequent Suzuki-Miyaura cross-coupling reaction, a silylcupration on a nonterminal acetylene, and an iododesilylation were the key steps in a convergent total synthesis of (-)-ebelactone A.
Low-molecular-weight immunomodifiers produced by micro-organisms.
H Umezawa
Biotechnology & genetic engineering reviews, 3, 255-273 (1985-01-01)
A Scaloni et al.
The Journal of biological chemistry, 269(21), 15076-15084 (1994-05-27)
The presence of a cysteine residue(s) near the active site of acylpeptide hydrolase was suggested by inactivation of the enzyme with sulfhydryl-modifying agents and by the substantial protection against inactivation afforded by the competitive inhibitor acetylmethionine. 5,5'-dithiobis-(2-nitrobenzoate) titrations of the
R Senthilkumar et al.
Experimental eye research, 72(3), 301-310 (2001-02-22)
Acylpeptide hydrolase removes the N -acetylated amino acids from the peptide substrates but not from intact proteins. Cleavage between amino acid residues 203--204 of the native acylpeptide hydrolase results in the formation of a 55 kDa truncated active enzyme in
Gianfranco De Pascale et al.
The Journal of antibiotics, 64(7), 483-487 (2011-04-28)
Homoserine transacetylase (HTA) catalyzes the transfer of an acetyl group from acetyl-CoA to the hydroxyl group of homoserine. This is the first committed step in the biosynthesis of methionine (Met) from aspartic acid in many fungi, Gram-positive and some Gram-negative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service