Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E008906

Sigma-Aldrich

Dextran sulfate sodium salt from Leuconostoc spp.

average Mw >500,000 (dextran starting material), contains 0.5-2% phosphate buffer

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

grade

Molecular Biology

form

powder

mol wt

average Mw >500,000 (dextran starting material)

contains

0.5-2% phosphate buffer

availability

available only in China

solubility

H2O: 100 mg/mL

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service