Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DIF001

Sigma-Aldrich

3T3-L1 DIFFERENTIATION KIT

Sufficient for 100 mL differentiation medium

Synonym(s):

3T3-L1 Differentiation Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

form

solution

usage

100 mL differentiation medium

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

storage temp.

−20°C

General description

3T3-L1 cells are derived from mouse 3T3 cells and provide a widely-used fundamental model in the study of adipose physiology and metabolic diseases. They exhibit a fibroblast-like morphology before differentiation but become rounded and accumulate lipid droplets several days after the initiation of differentiation. The accumulated lipid droplets can be visualized by light microscopy. The in vitro differentiated 3T3-L1 adipocytes result in characteristics similar to tissue-derived adipocytes and have been commonly used to study adipogenesis, lipolysis, and metabolic dysfunctions. The 3T3-L1 Differentiation Kit provides enough supplements to make 100 ml of differentiation medium and 600 ml of maintenance medium which is sufficient material for 12 100 mm culture dishes.

Application

3T3-L1 DIFFERENTIATION KIT has been used in:
  • Differentiation of 3T3-L1 preadipocytes to adipocytes.[1]
  • Study of obesity, adipogenesis, lipolysis and lipid metabolism
  • Differentiation of 3T3-L1 preadipocytes to adipocytes
  • Study of obesity, adipogenesis, lipolysis and lipid metabolism.

Components

Insulin (1.5 mg/ml) 0.6 ml (Catalog Number DIF001A, Green Cap)
Differentiation Cocktail, 1000x (lyophilized) 1 vl (Catalog Number DIF001B, Yellow Cap)
DMSO (anhydrous) 0.5 ml (Catalog Number DIF001C, Blue Cap)

Preparation Note

Insulin (1.5 mg/ml) 0.6 ml (Catalog Number DIF001A, Green Cap)
Differentiation Cocktail, 1000x (lyophilized) 1 vl (Catalog Number DIF001B, Yellow Cap)
DMSO (anhydrous) 0.5 ml (Catalog Number DIF001C, Blue Cap)

Storage and Stability

Store the kit at −20 °C, protected from light.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jérémie Gautheron et al.
Nature communications, 7, 11869-11869 (2016-06-22)
Receptor-interacting protein kinase 3 (RIPK3) mediates necroptosis, a form of programmed cell death that promotes inflammation in various pathological conditions, suggesting that it might be a privileged pharmacological target. However, its function in glucose homeostasis and obesity has been unknown.
Fuu-Jen Tsai et al.
Frontiers in pharmacology, 9, 1004-1004 (2018-09-21)
Antiretroviral (ART) drugs has previously been associated with lipodystrophic syndrome, metabolic consequences, and neuropsychiatric complications. ART drugs include three main classes of protease inhibitors (PIs), nucleoside analog reverse transcriptase inhibitors (NRTIs), and non-nucleoside reverse transcriptase inhibitors (NNRTIs). Our previous work
Tamar Golan et al.
Science signaling, 12(591) (2019-07-25)
Transforming growth factor-β (TGF-β) superfamily members are critical signals in tissue homeostasis and pathogenesis. Melanoma grows in the epidermis and invades the dermis before metastasizing. This disease progression is accompanied by increased sensitivity to microenvironmental TGF-β. Here, we found that
Ke Song et al.
JCI insight, 5(21) (2020-10-02)
Chronic kidney disease (CKD) induces the failure of arteriovenous fistulas (AVFs) and promotes the differentiation of vascular adventitial GLI1-positive mesenchymal stem cells (GMCs). However, the roles of GMCs in forming neointima in AVFs remain unknown. GMCs isolated from CKD mice

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service