Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D9267

Supelco

Dihydroouabain

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H46O12
CAS Number:
Molecular Weight:
586.67
EC Number:
MDL number:
UNSPSC Code:
41116107

SMILES string

[H][C@@]12CC[C@]3(O)C[C@H](C[C@@H](O)[C@]3(CO)[C@@]1([H])[C@H](O)C[C@]4(C)[C@H](CC[C@]24O)[C@H]5COC(=O)C5)O[C@@H]6O[C@@H](C)[C@H](O)[C@@H](O)[C@H]6O

InChI

1S/C29H46O12/c1-13-22(34)23(35)24(36)25(40-13)41-15-8-19(32)28(12-30)21-17(3-5-27(28,37)9-15)29(38)6-4-16(14-7-20(33)39-11-14)26(29,2)10-18(21)31/h13-19,21-25,30-32,34-38H,3-12H2,1-2H3/t13-,14?,15-,16+,17+,18+,19+,21+,22-,23+,24+,25-,26+,27-,28+,29-/m0/s1

InChI key

ZTFGOPUOTATSAL-CFVDWFIMSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F V Bielen et al.
Naunyn-Schmiedeberg's archives of pharmacology, 345(1), 100-107 (1992-01-01)
The kinetics of the inhibition by dihydroouabain (DHO) of the Na pump current were studied in isolated rabbit cardiac Purkinje cells by means of whole-cell recording. A fast exchange of the extracellular solution at the membrane of the cell studied
D J Mogul et al.
Circulation research, 64(6), 1063-1069 (1989-06-01)
Binding of cardiac glycosides to the Na+,K+-dependent ATPase has been shown to occur at both high- and low-affinity sites. However, recent reports suggest that glycoside-induced inhibition of electrogenic Na-K pump current occurs with simple first-order binding kinetics at relatively low-affinity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service