Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D9160

Sigma-Aldrich

1-Deoxymannojirimycin hydrochloride

Synonym(s):

1-Deoxymannojirimycin HCl, 1,5-Dideoxy-1,5-imino-D-mannitol hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$221.00
5 MG
$503.00
10 MG
$978.00

$221.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$221.00
5 MG
$503.00
10 MG
$978.00

About This Item

Empirical Formula (Hill Notation):
C6H13NO4 · HCl
CAS Number:
Molecular Weight:
199.63
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.76

$221.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

color

white to off-white

solubility

H2O: 10 mg/mL

antibiotic activity spectrum

viruses

Mode of action

enzyme | inhibits

storage temp.

2-8°C

SMILES string

Cl.OC[C@H]1NC[C@@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H13NO4.ClH/c8-2-3-5(10)6(11)4(9)1-7-3;/h3-11H,1-2H2;1H/t3-,4-,5-,6-;/m1./s1

InChI key

ZJIHMALTJRDNQI-MVNLRXSJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: glucosamine

Application

1-Deoxymannojirimycin (DMM) hydrochloride is an inhibitor of N-linked glycosylation and has antiviral activity. Derivatives of deoxymannojirimycin may have anti-HIV activity. It has been used for treatment of T lymphocytes and to study it′s effect on allogeneic cytotoxic T lymphocytes (CTL)[1]. It is used for studies on Golgi-mediated glycoprotein processing.

Biochem/physiol Actions

1-Deoxymannojirimycin is a mannosidase I inhibitor, alters cell-surface complex carbohydrate structure as assayed by PHA-L binding, and increases high mannose structures as indicated by the increased Con-A binding[1].
Selectively inhibits Golgi associated α(1→2) mannosidase and for studies on Golgi-mediated glycoprotein processing.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E U Bagriaçik et al.
Glycobiology, 6(4), 413-421 (1996-06-01)
The question whether or not target cell N-linked carbohydrate participates in cytotoxic T lymphocyte (CTL) mediated cytolysis has been in contention for well over a decade. Much of the evidence supporting a role for N-linked carbohydrate stems from the treatment
Janike Ehret et al.
Biotechnology and bioengineering, 116(4), 816-830 (2018-12-16)
Glycosylation is a key critical quality attribute for monoclonal antibodies and other recombinant proteins because of its impact on effector mechanisms and half-life. In this study, a variety of compounds were evaluated for their ability to modulate glycosylation profiles of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service