Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D8882

Sigma-Aldrich

Diphenylamine

crystalline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2NH
CAS Number:
Molecular Weight:
169.22
Beilstein:
508755
EC Number:
MDL number:
UNSPSC Code:
12352103

vapor density

5.82 (vs air)

vapor pressure

1 mmHg ( 108 °C)

form

crystalline

autoignition temp.

1175 °F

color

tan

bp

302 °C (lit.)

mp

50-53 °C (lit.)

SMILES string

N(c1ccccc1)c2ccccc2

InChI

1S/C12H11N/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h1-10,13H

InChI key

DMBHHRLKUKUOEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Target Organs

Kidney,Liver,spleen

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul Bamborough et al.
Journal of medicinal chemistry, 54(14), 5131-5143 (2011-06-28)
A kinase-focused screening set of fragments has been assembled and has proved successful for the discovery of ligand-efficient hits against many targets. Here we present some of our general conclusions from this exercise. Notably, we present the first profiling results
You-Zhi Tang et al.
Bioorganic & medicinal chemistry, 15(5), 1903-1913 (2007-01-24)
Since the research on antioxidants provides theoretical information for the medicinal development, and supplies some in vitro methods for quick-optimizing drugs, it attracts more scientific attention from bioorganic and medicinal chemists. In addition to the traditional O-H bond-type antioxidant, carbazole
Masaaki Fujikawa et al.
Bioorganic & medicinal chemistry, 15(11), 3756-3767 (2007-04-10)
We previously reported a classical quantitative structure-activity relationship (QSAR) equation for permeability coefficients (P(app-pampa)) by parallel artificial membrane permeation assay (PAMPA) of structurally diverse compounds with simple physicochemical parameters, hydrophobicity at a particular pH (logP(oct) and |pK(a)-pH|), hydrogen-accepting ability (SA(HA))

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service