Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D8696

Sigma-Aldrich

DMH4

>99.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H24 N4O2
CAS Number:
Molecular Weight:
400.47
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

>99.0% (HPLC)

form

powder

storage condition

desiccated

color

off-white to brown

solubility

DMSO: 20 mg/mL

originator

Merck & Co., Inc., Kenilworth, NJ, U.S.

storage temp.

2-8°C

SMILES string

C1CN(CCO1)CCOc2ccc(cc2)-c3cnc4c(cnn4c3)-c5ccccc5

InChI

1S/C24H24N4O2/c1-2-4-20(5-3-1)23-17-26-28-18-21(16-25-24(23)28)19-6-8-22(9-7-19)30-15-12-27-10-13-29-14-11-27/h1-9,16-18H,10-15H2

InChI key

SKZQZGSPYYHTQG-UHFFFAOYSA-N

Biochem/physiol Actions

DMH4 is a potent VEGF inhibitor and an angiogenesis inhbitor. It is a selective VEGF inhibitor with an IC50 of 161 nM for VEGFR inhibition compared to 8000 nM for AMPK. Unlike the structurally similar DMH1, which is a selective BMP inhibitor, DMH4 shows little affinitiy for BMP with an IC50 of 3500 nM for BMPR-I.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Merck & Co., Inc., Kenilworth, NJ, U.S.. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric M Erkenbrack et al.
Journal of experimental zoology. Part B, Molecular and developmental evolution, 328(5), 423-432 (2017-05-26)
Comparative studies of early development in echinoderms are revealing the tempo and mode of alterations to developmental gene regulatory networks and to the cell types they specify. In euechinoid sea urchins, skeletogenic mesenchyme (SM) ingresses prior to gastrulation at the
Zaheer Ali et al.
Arteriosclerosis, thrombosis, and vascular biology, 39(7), 1402-1418 (2019-06-27)
Objective- Pathological neovascularization is crucial for progression and morbidity of serious diseases such as cancer, diabetic retinopathy, and age-related macular degeneration. While mechanisms of ongoing pathological neovascularization have been extensively studied, the initiating pathological vascular remodeling (PVR) events, which precede
Sungwoon Lee et al.
The Journal of clinical investigation, 127(2), 457-471 (2016-12-20)
Controlled angiogenesis and lymphangiogenesis are essential for tissue development, function, and repair. However, aberrant neovascularization is an essential pathogenic mechanism in many human diseases, including diseases involving tumor growth and survival. Here, we have demonstrated that mice deficient in C-type
Liheng Shi et al.
Biochimica et biophysica acta, 1853(5), 1154-1164 (2015-02-24)
We previously identified peptide Lv, a novel bioactive peptide that enhances the activity of L-type voltage-gated calcium channels (L-VGCCs) in cone photoreceptors. In this study, we verified that peptide Lv was able to augment L-VGCC currents in cardiomyocytes, as well

Articles

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service