Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D8256

Sigma-Aldrich

Dansyl-DL-α-amino-n-butyric acid piperidinium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20N2O4S · C5H11N
CAS Number:
Molecular Weight:
421.55
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

−20°C

SMILES string

C1CCNCC1.CCC(NS(=O)(=O)c2cccc3c(cccc23)N(C)C)C(O)=O

InChI

1S/C16H20N2O4S.C5H11N/c1-4-13(16(19)20)17-23(21,22)15-10-6-7-11-12(15)8-5-9-14(11)18(2)3;1-2-4-6-5-3-1/h5-10,13,17H,4H2,1-3H3,(H,19,20);6H,1-5H2

InChI key

OHJSADPAHLBZEH-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service