Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D7297

Sigma-Aldrich

Deoxynucleotide Mix, 25 mM

Molecular Biology Reagent

Synonym(s):

25mM dNTP mix, dNTP mix, dNTPs

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 ML
$186.00
1 ML
$357.00

$186.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
0.5 ML
$186.00
1 ML
$357.00

About This Item

UNSPSC Code:
41106305
NACRES:
NA.21

$186.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

(PCR)

form

liquid

concentration

25 mM

color

colorless

shipped in

dry ice

storage temp.

−20°C

General description

The Deoxynucleotide mix is a convenient premixed dNTP solution containing 25 mM each of dATP, dCTP, dGTP, and TTP sodium salts in high-quality molecular biology grade water.

Application

Deoxynucleotide Mix, 25mM has been used as a component of the DNA amplification mixture for polymerase chain reaction (PCR).

Features and Benefits

  • Purity of each dNTP: Minimum 99%
  • Conveniently formulated; 1 μL is used per 50 μL PCR
  • Equimolar amounts of each dNTP means less pipetting
  • Minimize risk of contamination in PCR
  • UltraPure dNTPs can help maximize consistency and yields in critical PCR reactions

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service