Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D6908

Sigma-Aldrich

DL-erythro-Dihydrosphingosine

≥99%, synthetic

Synonym(s):

DL-erythro-1,3-Dihydroxy-2-aminooctadecane, DL-erythro-2-Amino-1,3-octadecanediol, DL-Sphinganine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H39NO2
CAS Number:
Molecular Weight:
301.51
Beilstein:
1724234
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
Pricing and availability is not currently available.

biological source

synthetic

Assay

≥99%

form

powder

solubility

chloroform/methanol (9:1): 20 mg/mL, clear, colorless to slightly yellow

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCC[C@H](O)[C@H](N)CO

InChI

1S/C18H39NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(21)17(19)16-20/h17-18,20-21H,2-16,19H2,1H3/t17-,18+/m1/s1

InChI key

OTKJDMGTUTTYMP-MSOLQXFVSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

D-Isomer is the biosynthetic precursor of sphingosine; inhibits protein kinase C, phospholipase A2, and phospholipase D.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Sakakura et al.
Biochemical and biophysical research communications, 246(3), 827-830 (1998-06-10)
Sphingosine (Sph) is emerging as an intracellular regulator of cellular differentiation and apoptosis (Ohta, et al., Cancer Res., 55, 691-697, 1995). We have recently found that both Sph and its methylated derivative N,N-dimethylsphingosine (DMS) inhibit mitogen-activated protein kinase (MAPK) activity
D D Archibald et al.
Biochimica et biophysica acta, 1166(2-3), 154-162 (1993-02-24)
The natural product D-erythro-sphingosine and synthetic racemic dihydrosphingosines were examined for their abilities to self-assemble into high-axial-ratio microstructures. When precipitated from methanol/water solution, D-erythro-sphingosine formed a viscoelastic gel composed of 50-nm diameter flexible fibers. These are 'cochleate cylinders' composed of
Emad Benlasher et al.
Avian diseases, 56(1), 120-127 (2012-05-02)
Fumonisins (FBs) are mycotoxins that are found worldwide in maize and maize products. Their main toxic effects have been well characterized in poultry, but differences between species have been demonstrated. Ducks appeared very sensitive to toxicity, whereas turkeys are more
Emma M Dangerfield et al.
Chembiochem : a European journal of chemical biology, 13(9), 1349-1356 (2012-05-29)
The immunomodulatory glycolipid α-galactosylceramide (α-GalCer) binds to CD1d and exhibits potent activity as a ligand for invariant CD1d-restricted natural killer-like T cells (iNKT cells). Structural analogues of α-GalCer have been synthesised to determine which components are required for CD1d presentation
Sang-Hun Song et al.
Scientific reports, 9(1), 9834-9834 (2019-07-10)
Surfactants during routine washing have a tremendous effect on lipid loss from hair. This study aims to understand the loss of lipids from hair upon contact with surfactants and develop a way to prevent the lipid loss. The change in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service