Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D6395

Sigma-Aldrich

trans-7, cis-9-Dodecadienyl acetate

Synonym(s):

(E,Z)-7,9-DDDA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H24O2
CAS Number:
Molecular Weight:
224.34
EC Number:
MDL number:
UNSPSC Code:
12352211

storage temp.

−20°C

SMILES string

CC/C=C\C=C\CCCCCCOC(C)=O

InChI

1S/C14H24O2/c1-3-4-5-6-7-8-9-10-11-12-13-16-14(2)15/h4-7H,3,8-13H2,1-2H3/b5-4-,7-6+

InChI key

LLRZUAWETKPZJO-SCFJQAPRSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Pheromone for Lobesia botrana

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service