Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D6389

Sigma-Aldrich

1,2-Didecanoyl-rac-glycerol

~99% (TLC)

Synonym(s):

1,2-Dicaprin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H44O5
CAS Number:
Molecular Weight:
400.59
MDL number:
UNSPSC Code:
12352211

Assay

~99% (TLC)

storage temp.

−20°C

SMILES string

CCCCCCCCCC(=O)OCC(CO)OC(=O)CCCCCCCCC

Biochem/physiol Actions

Cell-permeable activator of protein kinase C in platelets.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E G Lapetina et al.
The Journal of biological chemistry, 260(3), 1358-1361 (1985-02-10)
The ability of exogenous sn-1,2-diacylglycerols and analogs to function as bioregulators of protein kinase C in human platelets was investigated. The activation of protein kinase C in platelets is indicated by specific phosphorylation of a 40,000-dalton protein. Dihexanoylglycerol, dioctanoylglycerol (diC8)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service