Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D6068

Sigma-Aldrich

N-[4-[(6,7-Dimethoxy-4-quinazolinyl)amino]phenyl]benzamide hydrochloride

≥98% (HPLC), solid

Synonym(s):

4-(4′-Benzamidoanilino)-6,7-dimethoxyquinazoline hydrochloride, Aurora Kinase Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H20N4O3 · HCl
CAS Number:
Molecular Weight:
436.89
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

yellow

solubility

H2O: <2 mg/mL
DMSO: ~2.4 mg/mL (with warming up to 60°C for 1 minute)

originator

AstraZeneca

storage temp.

2-8°C

SMILES string

Cl.COc1cc2ncnc(Nc3ccc(NC(=O)c4ccccc4)cc3)c2cc1OC

InChI

1S/C23H20N4O3.ClH/c1-29-20-12-18-19(13-21(20)30-2)24-14-25-22(18)26-16-8-10-17(11-9-16)27-23(28)15-6-4-3-5-7-15;/h3-14H,1-2H3,(H,27,28)(H,24,25,26);1H

InChI key

OQCLKJREASBKNO-UHFFFAOYSA-N

Biochem/physiol Actions

Aurora Kinase Inhibitor

Features and Benefits

This compound was developed by AstraZeneca. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicola M Heron et al.
Bioorganic & medicinal chemistry letters, 16(5), 1320-1323 (2005-12-13)
A novel series of 5-aminopyrimidinyl quinazolines has been developed from anilino-quinazoline 1, which was identified in a high throughput screen for Aurora A. Introduction of the pyrimidine ring and optimisation of the substituents both on this ring and at the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service