Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

D5292

Sigma-Aldrich

DNA Ladder, Supercoiled

for supercoiled DNA electrophoresis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

grade

for molecular biology

form

liquid

usage

250 uses (25 μg size)
50 uses (10 μg size)

suitability

suitable for electrophoresis (DNA)

shipped in

dry ice

storage temp.

−20°C

General description

Sigma′s DNA Ladder (Supercoiled) contains 11 supercoiled DNA fragments from 2,067 – 16,210 bp. Ten ul of the ladder should be diluted in gel loading buffer and then loaded in a single lane on an agarose or polyacrylamide gel.

Components

Sigma′s DNA Ladder (Supercoiled) is provided in a solution of 10 mM Tris-HCl (pH 8.0), and 1 mM EDTA.

Other Notes

Contains 11 supercoiled fragments, 2,067–16,210 bp
For optimal resolution, the recommended agarose gel concentration is 0.7%.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael J Molloy et al.
Nucleic acids research, 32(16), e129-e129 (2004-09-11)
Within the biopharmaceutical industry, recombinant plasmid DNA is used both as a raw material (e.g. in lentiviral and AAV vector production) as well as an active ingredient (e.g. in DNA vaccines). Consequently, many analytical laboratories are routinely involved with plasmid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service