Skip to Content
MilliporeSigma
All Photos(1)

Documents

D5193

Sigma-Aldrich

17-Dimethylaminoethylamino-17-demethoxygeldanamycin

Synonym(s):

17-DMAG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H48N4O8
Molecular Weight:
616.75
UNSPSC Code:
51102829
NACRES:
NA.85

biological source

synthetic

Quality Level

form

powder

color

dark red-purple to black

solubility

ethanol: ~10 mg/mL (lit.)
DMSO: >25 mg/mL (lit.)

antibiotic activity spectrum

neoplastics

Mode of action

protein synthesis | interferes

storage temp.

−20°C

General description

Chemical structure: benzenoid

Application

17-DMAG has been used to block the NF-κB inflammatory cascade in the murine model. It a potential therapeutic agent for non-small-cell lung cancers (NSCLC).

Biochem/physiol Actions

17-DMAG is a heat shock protein (HSP90) inhibitor. 17-DMAG is a more potent analog of geldanamycin. It inhibits cancer growth and promotes apoptosis in multiple cell lines. 17-DMAG is a more potent antitumor agent than 17-AAG.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naruyuki Kobayashi et al.
Lung cancer (Amsterdam, Netherlands), 75(2), 161-166 (2011-07-20)
Acquired resistance to epidermal growth factor receptor-tyrosine kinase inhibitors (EGFR-TKIs), gefitinib and erlotinib, is frequently observed after initiation of TKIs therapy. Non-small-cell lung cancers (NSCLC) with activating EGFR mutations were reported to be sensitive to heat shock protein 90 (Hsp90)
HSP90 inhibition by 17-DMAG reduces inflammation in J774 macrophages through suppression of Akt and nuclear factor-κB pathways.
Shimp SK 3rd, Parson CD, et al.
Inflammation Research, epub (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service