Skip to Content
MilliporeSigma
All Photos(1)

Documents

D4774

Sigma-Aldrich

Dynorphin A Porcine Fragment 1-10

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H91N19O12
CAS Number:
Molecular Weight:
1234.45
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

storage temp.

−20°C

Amino Acid Sequence

Tyr-Gly-Gly-Phe-Leu-Arg-Arg-Ile-Arg-Pro

Biochem/physiol Actions

κ-agonist

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J E Morley et al.
Peptides, 4(6), 797-800 (1983-11-01)
Dynorphin-(1-17) produces a highly specific increase in food ingestion. Similar enhancement of food ingestion is found with dynorphin fragments (1-10), (1-11), (1-13) and (3-13) but not with (1-8) and (1-9). Dynorphin B (rimorphin) also enhances food intake. The highly specific

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service