Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D4634

Supelco

Dotriacontane

≥97% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)30CH3
CAS Number:
Molecular Weight:
450.87
Beilstein:
1783260
EC Number:
MDL number:
UNSPSC Code:
41116107

vapor density

15.6 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥97% (GC)

bp

467 °C (lit.)

mp

65-70 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C32H66/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-32H2,1-2H3

InChI key

QHMGJGNTMQDRQA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service