Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D4036

Sigma-Aldrich

Dynorphin A Porcine Fragment 1-9

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H84N18O11
CAS Number:
Molecular Weight:
1137.34
MDL number:
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](Cc1ccccc1)NC(=O)CNC(=O)CNC(=O)[C@@H](N)Cc2ccc(O)cc2)C(=O)N[C@@H](CCCNC(N)=N)C(O)=O

Gene Information

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Tyr-Gly-Gly-Phe-Leu-Arg-Arg-Ile-Arg

Biochem/physiol Actions

Potent κ-agonist

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Miller et al.
European journal of pharmacology, 116(1-2), 159-163 (1985-10-08)
The inhibitory action of dynorphin-(1-9) on the electrically stimulated mouse vas deferens was seen to be antagonised by the delta-selective opioid antagonist ICI 174864. The observed delta-receptor mediated responses were partially, but not totally, prevented by peptidase inhibitors which protect
M G Gillan et al.
Journal of neurochemistry, 45(4), 1034-1042 (1985-10-01)
Following incubation of [3H]dynorphin A (1-8) and [3H]dynorphin A (1-9) with suspensions of guinea pig brain membranes, analysis of the supernatants by HPLC has shown that both peptides are degraded at 25 degrees C and at 0 degrees C. Bestatin
M Maruyama et al.
Brain research, 401(1), 14-22 (1987-01-13)
This paper describes the binding properties of [3H]peptidergic opioid ligands to binding sites solubilized from rat brain membranes by the treatment with 0.125% sodium glycodeoxycholate and 1 M NaCl. The highest amount of the specific binding of [3H]-[D-Ala2-, Met5]enkephalinamide was
P Slater et al.
Neuropeptides, 8(1), 71-76 (1986-07-01)
The autoradiographic distribution of kappa opioid binding sites was evaluated in sections of monkey brain using the selective ligand [3H]dynorphin1-9. Kappa receptors were highly concentrated in the deep layers of the cerebral cortex, the substantia nigra, the hippocampus and the
M S Wood et al.
Neuropharmacology, 28(10), 1041-1046 (1989-10-01)
The binding of radiolabelled ligands with high affinity for kappa-opioid binding sites has been studied in homogenates of lumbo-sacral spinal cord from the rat. The unselective ligands [3H]bremazocine and [3H]diprenorphine labelled a large number of sites which could not be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service