Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D3413

Sigma-Aldrich

(5S,12S)-Dihydroxy-(6E,8E,10E,14Z)-eicosatetraenoic acid

≥98%, ~100 μg/mL in ethanol

Synonym(s):

5,12-DiHETE, (6E)-12-epi-Leukotriene B4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32O4
CAS Number:
Molecular Weight:
336.47
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

≥98%

form

liquid

concentration

~100 μg/mL in ethanol

functional group

carboxylic acid

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCC\C=C\CC(O)\C=C\C=C\C=C\C(O)CCCC(O)=O

InChI

1S/C20H32O4/c1-2-3-4-5-6-9-13-18(21)14-10-7-8-11-15-19(22)16-12-17-20(23)24/h6-11,14-15,18-19,21-22H,2-5,12-13,16-17H2,1H3,(H,23,24)/b8-7+,9-6+,14-10+,15-11+

InChI key

VNYSSYRCGWBHLG-XUOUMLBJSA-N

Packaging

Packaged under Argon.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup - Solvent

Flash Point(C)

14 °C - closed cup - Solvent

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service