Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D3409

Sigma-Aldrich

Diphenylamine

ACS reagent, passes test for sensitivity to nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2NH
CAS Number:
Molecular Weight:
169.22
Beilstein:
508755
EC Number:
MDL number:
UNSPSC Code:
12171500

grade

ACS reagent

vapor density

5.82 (vs air)

vapor pressure

1 mmHg ( 108 °C)

autoignition temp.

1175 °F

ign. residue

≤0.03%

bp

302 °C (lit.)

mp

50-53 °C (lit.)

solubility

alcohol: passes test

anion traces

nitrate (NO3-): passes test

suitability

passes test for sensitivity to nitrate

SMILES string

N(c1ccccc1)c2ccccc2

InChI

1S/C12H11N/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h1-10,13H

InChI key

DMBHHRLKUKUOEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Target Organs

Kidney,Liver,spleen

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul Bamborough et al.
Journal of medicinal chemistry, 54(14), 5131-5143 (2011-06-28)
A kinase-focused screening set of fragments has been assembled and has proved successful for the discovery of ligand-efficient hits against many targets. Here we present some of our general conclusions from this exercise. Notably, we present the first profiling results
You-Zhi Tang et al.
Bioorganic & medicinal chemistry, 15(5), 1903-1913 (2007-01-24)
Since the research on antioxidants provides theoretical information for the medicinal development, and supplies some in vitro methods for quick-optimizing drugs, it attracts more scientific attention from bioorganic and medicinal chemists. In addition to the traditional O-H bond-type antioxidant, carbazole
Masaaki Fujikawa et al.
Bioorganic & medicinal chemistry, 15(11), 3756-3767 (2007-04-10)
We previously reported a classical quantitative structure-activity relationship (QSAR) equation for permeability coefficients (P(app-pampa)) by parallel artificial membrane permeation assay (PAMPA) of structurally diverse compounds with simple physicochemical parameters, hydrophobicity at a particular pH (logP(oct) and |pK(a)-pH|), hydrogen-accepting ability (SA(HA))

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service