Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D3031

Sigma-Aldrich

Diethyl disulfide

Synonym(s):

Ethyl disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2S2
CAS Number:
Molecular Weight:
122.25
EC Number:
MDL number:
UNSPSC Code:
12352204

refractive index

n20/D 1.506 (lit.)

bp

151-153 °C (lit.)

density

0.993 g/mL at 25 °C (lit.)

SMILES string

CCSSCC

InChI

1S/C4H10S2/c1-3-5-6-4-2/h3-4H2,1-2H3

InChI key

CETBSQOFQKLHHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong-Qing Ye et al.
Food chemistry, 202, 236-246 (2016-02-28)
The evolution of volatile sulfur compounds (VSCs) in Cabernet Sauvignon wines from seven regions of China during maturation in oak barrels was investigated. The barrels were made of different wood grains (fine and medium) and toasting levels (light and medium).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service