Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D3022

Sigma-Aldrich

Diisopropylamine

≥99%

Synonym(s):

DIPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHNHCH(CH3)2
CAS Number:
Molecular Weight:
101.19
Beilstein:
605284
EC Number:
MDL number:
UNSPSC Code:
12352100

vapor density

3.5 (vs air)

vapor pressure

50 mmHg ( 20 °C)

Assay

≥99%

autoignition temp.

599 °F

expl. lim.

8.5 %

refractive index

n20/D 1.392 (lit.)

bp

84 °C (lit.)

mp

−61 °C (lit.)

density

0.722 g/mL at 25 °C (lit.)

SMILES string

CC(C)NC(C)C

InChI

1S/C6H15N/c1-5(2)7-6(3)4/h5-7H,1-4H3

InChI key

UAOMVDZJSHZZME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

7.8 °F - closed cup

Flash Point(C)

-13.45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Songping Liao et al.
Journal of the American Chemical Society, 125(49), 15114-15127 (2003-12-05)
Lithium diisopropylamide-mediated lithiations of N-alkyl ketimines derived from cyclohexanones reveal that simple substitutions on the N-alkyl side chain and the 2-position of the cyclohexyl moiety afford a 60,000-fold range of rates. Detailed rate studies implicate monosolvated monomers at the rate-limiting
Antonio Ramirez et al.
Journal of the American Chemical Society, 128(31), 10326-10336 (2006-08-03)
Structural, kinetic, and computational studies reveal the mechanistic complexities of a lithium diisopropylamide (LDA)-mediated ester enolization. Hemilabile amino ether MeOCH2CH2NMe2, binding as an eta1 (ether-bound) ligand in the reactant and as an eta2 (chelating) ligand in the transition structure, accelerates
Diisopropylamide and TMP turbo-Grignard reagents: a structural rationale for their contrasting reactivities.
David R Armstrong et al.
Angewandte Chemie (International ed. in English), 49(18), 3185-3188 (2010-03-31)
Kanwal Jit Singh et al.
Journal of the American Chemical Society, 128(42), 13753-13760 (2006-10-19)
Structural and mechanistic studies of the lithium diisopropylamide (LDA)-mediated anionic Fries rearrangements of aryl carbamates are described. Substituents at the meta position of the arene (H, OMe, F) and the dialkylamino moiety of the carbamate (Me(2)N, Et(2)N, and i-Pr(2)N) markedly
Mihai S Viciu et al.
Journal of the American Chemical Society, 132(18), 6361-6365 (2010-04-20)
Treatment of 2,6-difluoropyridine with lithium diisopropylamide in THF solution at -78 degrees C effects ortholithiation quantitatively. Warming the solution to 0 degrees C converts the aryllithium to 2-fluoro-6-(diisopropylamino)pyridine. Rate studies reveal evidence of a reversal of the ortholithiation and a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service