Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D2289

Sigma-Aldrich

Dermenkephalin

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H62N10O10S2
CAS Number:
Molecular Weight:
955.15
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

storage temp.

−20°C

Amino Acid Sequence

Tyr-D-Met-Phe-His-Leu-Met-Asp-NH2

Biochem/physiol Actions

Opioid peptide with selective affinity for δ-receptors

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Jhamandas et al.
Peptides, 27(5), 953-963 (2006-03-07)
Neuropeptide FF and related synthetic amidated peptides have been shown to elicit sustained anti-nociceptive responses and potently augment spinal anti-nociceptive actions of spinal morphine in tests of thermal and mechanical nociception. Recent studies have described the occurrence of another octapeptide
Deltorphin analogs restricted via a urea bridge: structure and opioid activity.
J Ciarkowski et al.
Advances in experimental medicine and biology, 611, 491-492 (2009-04-30)
Constance Auvynet et al.
European journal of cell biology, 85(1), 25-34 (2005-12-24)
The dermal glands of the South American hylid frog Phyllomedusa bicolor synthesize and expel huge amounts of cationic, alpha-helical, 24- to 33-residue antimicrobial peptides, the dermaseptins B. These glands also produce a wide array of peptides that are similar to
Anna Capasso
Current medicinal chemistry, 15(24), 2518-2526 (2008-10-16)
The possible role of PGs and NO in the development of S&W of DBA/2J mice was investigated by evaluating the effects of dexamethasone, indometacin, mifepristone plus dexamethasone on the S&W, as well as of L-NAME both on the S&W in
Kvar C Black et al.
Molecular imaging, 7(1), 50-57 (2008-04-04)
Molecularly targeted gold nanorods were investigated for applications in both diagnostic imaging and disease treatment with cellular resolution. The nanorods were tested in two genetically engineered cell lines derived from the human colon carcinoma HCT-116, a model for studying ligand-receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service