Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D1551

Sigma-Aldrich

Diethylenetriamine

≥99% (GC)

Synonym(s):

2,2′-Diaminodiethylamine, 2,2′-Iminodiethylamine, Bis(2-aminoethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH2CH2CH2)2NH
CAS Number:
Molecular Weight:
103.17
Beilstein:
605314
EC Number:
MDL number:
UNSPSC Code:
12352103

vapor density

3.6 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)

Assay

≥99% (GC)

autoignition temp.

676 °F

expl. lim.

6.7 %

refractive index

n20/D 1.484 (lit.)

bp

199-209 °C (lit.)

mp

−35 °C (lit.)

density

0.955 g/mL at 25 °C (lit.)

SMILES string

NCCNCCN

InChI

1S/C4H13N3/c5-1-3-7-4-2-6/h7H,1-6H2

InChI key

RPNUMPOLZDHAAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrizio Carta et al.
Journal of medicinal chemistry, 53(15), 5511-5522 (2010-07-02)
Carbonic anhydrases (CAs, EC 4.2.1.1) are inhibited by sulfonamides, phenols, and coumarins. Polyamines such as spermine, spermidine, and many synthetic congeners are described to constitute a novel class of CA inhibitors (CAIs), interacting with the different CA isozymes with efficiency

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service