Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D1301

Sigma-Aldrich

S-(4,5-Dimethoxy-2-nitrobenzyl)isothiouronium bromide

≥97%

Synonym(s):

2-(4,5-Dimethoxy-2-nitrobenzyl)-2-thiopseudourea hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H2(OCH3)2CH2SC(=NH)NH2 · HBr
CAS Number:
Molecular Weight:
352.20
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97%

mp

230-231 °C (dec.) (lit.)

SMILES string

Br[H].COc1cc(CSC(N)=N)c(cc1OC)[N+]([O-])=O

InChI

1S/C10H13N3O4S.BrH/c1-16-8-3-6(5-18-10(11)12)7(13(14)15)4-9(8)17-2;/h3-4H,5H2,1-2H3,(H3,11,12);1H

InChI key

LOQPMIGKYIVTQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service