Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D124

Supelco

Doxepin hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21NO · HCl
CAS Number:
Molecular Weight:
315.84
EC Number:
MDL number:
UNSPSC Code:
41116107

SMILES string

Cl.CN(C)CC\C=C1/c2ccccc2COc3ccccc13

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julia Rossmann et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1000, 84-94 (2015-07-27)
Matrix effects that occur during quantitative measurement by liquid chromatography mass spectrometry specifically when using electrospray ionization are a widely recognized phenomenon. Sample matrix compounds affect the ionization process of the target analytes, lead to a low signal response, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service