Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

D0947

Sigma-Aldrich

Dihydrotanshinone I

≥98% (HPLC)

Synonym(s):

Dihydrotanshinone I, (-)-Dihydrotanshinone I, 15,16-Dihydrotanshinone I

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$245.00
50 MG
$786.00

$245.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$245.00
50 MG
$786.00

About This Item

Empirical Formula (Hill Notation):
C18H14O3
CAS Number:
Molecular Weight:
278.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

$245.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

Salvia miltiorrhiza

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

red

solubility

ethanol: 1 mg/mL, clear, orange to red

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

C[C@H]1COC(C2=C3C4=C(C=C2)C(C)=CC=C4)=C1C(C3=O)=O

InChI

1S/C18H14O3/c1-9-4-3-5-12-11(9)6-7-13-15(12)17(20)16(19)14-10(2)8-21-18(13)14/h3-7,10H,8H2,1-2H3/t10-/m0/s1

InChI key

HARGZZNYNSYSGJ-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

General description

DihydrotanshinoneI (DHTS) is a tanshinone derivative which is a soluble diterpene quinone pigment of danshen.

Application

Dihydrotanshinone I (DHTS) has been used:
  • to study its cytocidal effects on chemosensitive ovarian cancer cells with or without platinum-based chemotherapy
  • as a treatment to study its inhibitory effects on triple-negative breast cancer cells growth by modulating epithelial-mesenchymal transition (EMT) markers
  • as a potential SARS-CoV-2 papain-like protease (PLpro) inhibitor
  • as a human antigen R (HuR) inhibitor to study its role in Th17 cell differentiation related to autoimmune neuroinflammation
  • as a reference standard to analyze Salvia miltiorrhiza Bunge extract (SME) components using high-performance liquid chromatography(HPLC)

Biochem/physiol Actions

Dihydrotanshinone Iis a potent anti-cancer agent with its cytotoxic, apoptosis, cell cycle arresteffects and inhibition of angiogenesis, and metastasis in cancer cells. Italso exerts cardiovascular and cerebrovascular protective effects. DHTS isan anti-inflammatory, anti-ischemia-reperfusion (I/R), anti-allergic,anti-Alzheimer’s disease properties. It is also effective againstdrug-resistant cancer cells.
Tanshinone extracted from a herb, Salvia miltiorrhiza, commonly used in Chinese traditional medicine. A significant decrease in tumor growth was observed using dihydrotanshinone I and irradiation treatment with mouse xenograft model.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W D Lee et al.
Nanotechnology, 27(49), 495101-495101 (2016-11-12)
Danshen (Salvia miltiorrhiza), a Chinese medicinal herb, consists of several functional components including tanshinones responsible for prevention of several chronic diseases. This study intends to prepare tanshinone extract and nanoemulsion from danshen and determine their inhibition effect on lung cancer
Sarah R Anthony et al.
Adipocyte, 9(1), 335-345 (2020-07-28)
The goal of this study was to define the functional role of adipocyte-specific expression of the RNA binding protein Human antigen R (HuR). Mice with an adipocyte-specific deletion of HuR (Adipo-HuR-/- ) were generated by crossing HuR floxed (HuRfl/fl )
Jiliang Cao et al.
Journal of chromatographic science, 54(8), 1435-1444 (2016-05-04)
A simple and fast micellar electrokinetic chromatography (MEKC) method using ionic liquids as modifier was established for simultaneous determination of four hydrophobic tanshinones, including dihydrotanshinone I ( 1: ), cryptotanshinone ( 2: ), tanshinone I ( 3: ) and tanshinone
Akanksha Kashyap et al.
Asian Pacific journal of cancer prevention : APJCP, 22(7), 2177-2184 (2021-07-29)
Salvia miltiorrhiza Bunge (Danshen), has been used for its therapeutic value in Traditional Chinese Medicine (TCM), for almost a thousand years. Dihydrotanshinone-I (DHTS) is a lipophilic compound isolated from the plant Salvia miltiorrhiza that has been shown to induce anti-proliferative
Ok Kyu Park et al.
Fitoterapia, 83(8), 1666-1674 (2012-10-02)
We observed neuroprotective effects of five major lipophilic diterpenes derived from Danshen (Radix Salvia miltiorrhiza) extract, such as cryptotanshinone (CTs), dihydrotanshinone I (DTsI), tanshinone I (TsI), tanshinone IIA (TsIIA) and tanshinone IIB (TsIIB), in the hippocampal CA1 region (CA1) against

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service