Skip to Content
MilliporeSigma
All Photos(1)

Documents

D0890

Sigma-Aldrich

1,2:3,5-Di-O-isopropylidene-α-D-apiose

Synonym(s):

α-D-Apiose diacetonide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H18O5
CAS Number:
Molecular Weight:
230.26
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

CC1(C)OCC2(COC3OC(C)(C)OC23)O1

InChI

1S/C11H18O5/c1-9(2)13-6-11(16-9)5-12-8-7(11)14-10(3,4)15-8/h7-8H,5-6H2,1-4H3

InChI key

WGCOBUGUSFKJSL-UHFFFAOYSA-N

Application

Di-O-isopropylidene-α-D-apiose may be used as a reagent for the organic synthesis of conformationally semirigid diacylglycerol (DAG) analogues embedded into five-membered ring lactones.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Lee et al.
Journal of medicinal chemistry, 39(25), 4912-4919 (1996-12-06)
In the present investigation, the last two possible modes of generating conformationally semirigid diacylglycerol (DAG) analogues embedded into five-membered ring lactones as templates III and IV are investigated. The first two templates studied in previous investigations corresponded to 2-deoxyribonolactone (template

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service