Skip to Content
MilliporeSigma
All Photos(1)

Documents

D0758

Sigma-Aldrich

2′-Deoxyinosine 5′-triphosphate trisodium salt

synthetic, 95-97%

Synonym(s):

dITP-Na3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N4Na3O13P3
CAS Number:
Molecular Weight:
558.11
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

biological source

synthetic

Assay

95-97%

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].O[C@H]1C[C@@H](O[C@@H]1COP([O-])(=O)OP([O-])(=O)OP(O)([O-])=O)n2cnc3C(=O)NC=Nc23

InChI

1S/C10H15N4O13P3.3Na/c15-5-1-7(14-4-13-8-9(14)11-3-12-10(8)16)25-6(5)2-24-29(20,21)27-30(22,23)26-28(17,18)19;;;/h3-7,15H,1-2H2,(H,20,21)(H,22,23)(H,11,12,16)(H2,17,18,19);;;/q;3*+1/p-3/t5-,6+,7+;;;/m0.../s1

InChI key

FHYSVBOUQFJKCI-PWDLANNDSA-K

Looking for similar products? Visit Product Comparison Guide

Application

2′-Deoxyinosine 5-triphosphate (dITP) may be formed by nitrosative deamination processes such as those that deaminate adenosine in DNA and RNA. dITP may be useful in studies of this process.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Auer et al.
Nucleic acids research, 24(24), 5021-5025 (1996-12-15)
The ability to selectively amplify RNA in the presence of genomic DNA of analogous sequence is cumbersome and requires implementation of critical controls for genes lacking introns. The convenient approaches of either designing oligonucleotide primers at the splice junction or
Gel compressions and artifact banding can be resolved in the same DNA sequence reaction.
K W McCrea et al.
BioTechniques, 15(5), 843-844 (1993-11-01)
ITPA protein, an enzyme that eliminates deaminated purine nucleoside triphosphates in cells.
Sakumi K, Abolhassani N, et al.
Mutation Research, 703, 4-50 (2010)
Anatoli Tchigvintsev et al.
The Biochemical journal, 437(2), 243-253 (2011-05-10)
Genomes of all free-living organisms encode the enzyme dUTPase (dUTP pyrophosphatase), which plays a key role in preventing uracil incorporation into DNA. In the present paper, we describe the biochemical and structural characterization of DUT1 (Saccharomyces cerevisiae dUTPase). The hydrolysis
Yoshihito Ueno et al.
Bioorganic & medicinal chemistry, 12(24), 6581-6586 (2004-11-24)
In this paper, we describe the synthesis and thermal stabilities of the triplexes containing either 2'-deoxyinosine (1) or 2'-deoxyxanthosine (3) in their second strands. It was found that the triplexes with the 2'-deoxy-5-methylcytidine(dM)*1:dC and dM*1:dA base triplets are thermally stable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service