Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D0506

Sigma-Aldrich

Dansyl-L-α-amino-n-butyric acid cyclohexylammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C16H20N2SO4 · C6H13N
CAS Number:
Molecular Weight:
435.58
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

NC1CCCCC1.CC[C@H](NS(=O)(=O)c2cccc3c(cccc23)N(C)C)C(O)=O

InChI

1S/C16H20N2O4S.C6H13N/c1-4-13(16(19)20)17-23(21,22)15-10-6-7-11-12(15)8-5-9-14(11)18(2)3;7-6-4-2-1-3-5-6/h5-10,13,17H,4H2,1-3H3,(H,19,20);6H,1-5,7H2/t13-;/m0./s1

InChI key

GPDKVEDZOSFQHF-ZOWNYOTGSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service