Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D0296

Sigma-Aldrich

N-(2,4-Dinitrophenyl)-Pro-Leu-Gly-Cys(Me)-His-Ala-D-Arg amide

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H57N15O11S
CAS Number:
Molecular Weight:
932.02
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

Assay

≥95% (HPLC)

storage temp.

−20°C

SMILES string

CSCC(NC(=O)CNC(=O)C(CC(C)C)NC(=O)C1CCCN1c2ccc(cc2N(=O)=O)N(=O)=O)C(=O)NC(Cc3cnc[nH]3)C(=O)NC(C)C(=O)NC(CCCNC(N)=N)C(N)=O

Substrates

An "optimized" fluorogenic substrate for collagenase.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Berman et al.
The Journal of biological chemistry, 267(3), 1434-1437 (1992-01-25)
A strategy is described for the rapid optimization of kcat/Km for protease substrates. Selected positions of a given peptide substrate sequence are varied through synthesis with mixtures of amino acids. Incubation of the resulting peptide mixture with the enzyme of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service