Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D0255

Sigma-Aldrich

N,N′-Di(2,4-dinitrophenyl)-L-lysine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18N6O10
CAS Number:
Molecular Weight:
478.37
EC Number:
UNSPSC Code:
12352200

storage temp.

−20°C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R E Kleinman et al.
Immunology, 68(4), 464-468 (1989-12-01)
We examined the mucosal barrier function during anaphylaxis induced by the hapten N,N'-di-2,4,dinitrophenyl-lysine (di-DNP-lysine) in BDF1 female mice immunized with dinitrophenylated Ascaris suum extract. Immunized mice were gavaged with 10 mg or 50 mg of ovalbumin (OVA) with or without
A Leonardi et al.
Ophthalmic research, 23(4), 187-195 (1991-01-01)
Clinical and histological profiles of guinea pig conjunctiva undergoing late-phase reaction (LPR) were evaluated before and after additional antigenic challenge. Four groups of animals were passively sensitized with IgG1 antibodies, challenged with the specific hapten di-DNP-lysine, and rechallenged during LPR
Obligatory hybridization of heterologous immunoglobulin light chains into covalently linked dimers.
D S Peabody et al.
Biochemistry, 19(13), 2827-2834 (1980-06-24)
S D Trocmé et al.
Current eye research, 7(5), 437-443 (1988-05-01)
A cellular late-phase reaction is described in a rat model of topically induced ocular anaphylaxis. Rats were immunized with dinitrophenylated Ascaris suum extract and alum and were tested for active cutaneous anaphylaxis on day 13. Rats with a strong skin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service