Skip to Content
MilliporeSigma

Skip To

D014

6-Hydroxydopamine hydrobromide

Synonym(s):

2,4,5-Trihydroxyphenethylamine hydrobromide, 2,5-Dihydroxytyramine hydrobromide, 2-(2,4,5-Trihydroxyphenyl)ethylamine hydrobromide, 6-OHDA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
(HO)3C6H2CH2CH2NH2 · HBr
CAS Number:
Molecular Weight:
250.09
EC Number:
211-247-0
UNSPSC Code:
41116107
MDL number:
Beilstein/REAXYS Number:
3713280
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


mp

216-220 °C (lit.)

storage temp.

−20°C

SMILES string

Br.NCCc1cc(O)c(O)cc1O

InChI

1S/C8H11NO3.BrH/c9-2-1-5-3-7(11)8(12)4-6(5)10;/h3-4,10-12H,1-2,9H2;1H

InChI key

MLACDGUOKDOLGC-UHFFFAOYSA-N

General description

Solutions should be freshly prepared and protected from exposure to light.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library