Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

CWA-1085

Sigma-Aldrich

Anti-Mouse CD16/CD32 (93) ColorWheel® Dye-Ready mAb

greener alternative

for use with ColorWheel® Dyes (Required, (Sold Separately))

Synonym(s):

Low affinity immunoglobulin gamma Fc region receptor III/II

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
93, monoclonal
application:
FACS
species reactivity:
mouse
technique(s):
flow cytometry: suitable
citations:

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

93, monoclonal

product line

ColorWheel®

form

lyophilized

mol wt

calculated mol wt 30.04 and 36.95 kDa

purified by

using Protein A

species reactivity

mouse

species reactivity (predicted by homology)

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable

isotype

IgG2aκ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

compatibility

for use with ColorWheel® Dyes (Required, (Sold Separately))

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

ColorWheel® technology is a novel and proprietary method of creating your own antibody and dye combinations for use in flow cytometry. By incubating any ColorWheel® antibody with any ColorWheel® dye, researchers can quickly and simply produce primary conjugated antibodies for use in single color or multicolor flow cytometry analysis. Each ColorWheel® antibody and dye is lyophilized for long-term storage, allowing for simplicity and flexibility without compromise. ColorWheel® technology requires both ColorWheel® antibody and ColorWheel® dye for Flow Cytometry application.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This product is Preservative-free, lyophilized product for enhanced stability and allow for ambient shipping and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Specificity

Clone 93 is a rat monoclonal antibody that detects CD16/CD32. It targets a conformational epitope formed by FcgII and FcgIII receptors.

Immunogen

VH81X Tg B cells and T220 stromal cells.

Application

Quality Control Testing

Evaluated by Flow Cytometry in BALB/c mouse splenocytes.

Flow Cytometry Analysis: Staining of one million BALB/c mouse splenocytes was performed using 5 μL of a 1:1 mixture of Cat. No. CWA-1085, Anti-Mouse CD16/CD32 (93) ColorWheel® Dye-Ready mAb and Cat. No. CWDS-PE ColorWheel® Antibody-Ready Phycoerythrin (PE) Dye or an equivalent amount of PE-conjugated Rat IgG2a isotype control.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Compatibility

Requires ColorWheel® Dye (Sold Separately)

Target description

Low affinity immunoglobulin gamma Fc region receptor III/II (UniProt: P08508/P08101; also known as IgG Fc receptor III/II, FcRIII/II, CD16/CD32) are encoded by the Fcgr3/Fcgr2 genes (Gene ID: 14131/14130) in murine species. CD16 and CD32 are single-pass type I membrane glycoproteins that are synthesized with a signal peptide (aa 1-30 and 1-29), which is subsequently cleaved off to produce the mature form that contains an extracellular domain (aa 31-215 and 30-210), a transmembrane domain (aa 216-235 and 211-231), and a cytoplasmic domain (aa 236-261 and 232-330). CD16/CD32 are expressed on B cells, monocytes/macrophages, NK cells, granulocytes, mast cells, and dendritic cells. Both CD16 and CD32 can be expressed on the same cell and in varying ratios. They are constitutively expressed, although cytokines and lymphokines can modulate their expression. They serve as low-affinity receptors and bind to the Fc portion of antibody-antigen complexes and play a role in adaptive immune responses. They are involved in a variety of effector and regulatory functions, including phagocytosis of antigen-antibody complexes from the circulation and modulation of antibody production by B-cells. Clone 93 recognizes the common epitope on CD16/CD32.

Physical form

Lyophilized from PBS containing D-Mannitol and Sucrose, normal appearance is a dried pellet. Reconstituted antibody solution is stable and functional as assessed by functional testing. Contains no biocide or preservatives, such as azide.

Reconstitution

1.0 mg/mL after reconstitution at 25 μL in PBS. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C. Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial. Reconstitute each vial by adding 25 μL PBS. Protect from light.

Legal Information

ColorWheel is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

required but not provided

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service