Skip to Content
MilliporeSigma
All Photos(7)

Documents

CWA-1035

Sigma-Aldrich

Anti-Human CD8a (RPA-T8) ColorWheel® Dye-Ready mAb

greener alternative

for use with ColorWheel® Dyes (Required, sold separately)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

RPA-T8, monoclonal

product line

ColorWheel®

form

lyophilized

mol wt

calculated mol wt 25.73 kDa

species reactivity

human

species reactivity (predicted by homology)

nonhuman primates

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable

isotype

IgG1κ

epitope sequence

Unknown.

Protein ID accession no.

UniProt accession no.

compatibility

for use with ColorWheel® Dyes (Required, sold separately)

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD8A;MAL(925)

General description

ColorWheel® technology is a novel and proprietary method of creating your own antibody and dye combinations for use in flow cytometry. By incubating any ColorWheel® antibody with any ColorWheel® dye, researchers can quickly and simply produce primary conjugated antibodies for use in single color or multicolor flow cytometry analysis. Each ColorWheel® antibody and dye is lyophilized for long long-term storage, allowing for simplicity and flexibility without compromise. ColorWheel® technology requires both ColorWheel® antibody and ColorWheel® dye for Flow Cytometry application.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This product is Preservative-free, lyophilized product for enhanced stability and allow for ambient shipping and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Specificity

Clone RPA-T8 is a mouse monoclonal antibody that detects human T-cell surface glycoprotein CD8 alpha chain (CD8a).

Application

Quality Control Testing:

Evaluated by Flow Cytometry in human peripheral blood mononuclear cells (PBMC).

 Staining of one million Human peripheral blood mononuclear cells (PBMC) was performed using 5 μL of a 1:1 mixture of Cat. No. CWA-1035, Anti-Human CD8a (RPA-T8) ColorWheel® Dye-Ready mAb and Cat. No. CWD-PE ColorWheel® Antibody-Ready Phycoerythrin (PE) Dye or an equivalent amount of PE-conjugated Mouse IgG1 isotype control.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Compatibility

Requires ColorWheel® Dye (Sold Separately)

Target description

T-cell surface glycoprotein CD8 alpha chain (UniProt: P01732; also known as T-lymphocyte differentiation antigen T8/Leu-2, CD8a) is encoded by the CD8A (also known as MAL) gene (Gene ID: 925) in human. CD8 alpha is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-21), which is subsequently cleaved to form the mature protein that has an extracellular domain (aa 22-182), a helical domain (aa 183-203), and a cytoplasmic tail (aa 204-235). CD8 is generally present as a heterodimer of alpha and beta chains linked by two disulfide bonds, however, it can also form homodimers. The CD8 alpha/alpha homodimer is expressed on a subset of gamma/delta TCR-bearing T cells, NK cells, intestinal intraepithelial lymphocytes, and lymphoid dendritic cells. CD8a homodimer is also reported to promote the survival and differentiation of activated lymphocytes into memory CD8 T-cells. CD8a plays an essential role in the immune response and serves multiple functions in responses against both external and internal offenses. In T-cells, it functions primarily as a coreceptor for MHC class I molecule:peptide complex. It interacts simultaneously with the T-cell receptor (TCR) and the MHC class I proteins presented by antigen presenting cells. In turn, it recruits the Src kinase LCK to the vicinity of the TCR-CD3 complex where LCK phosphorylates various substrates leading to lymphokine production, motility, adhesion, and activation of cytotoxic T-lymphocytes (CTLs). This process enables CTLs to recognize and eliminate infected cells and tumor cells. (Ref.: Harland, KL., et al., (2014). Nat. Commun. 5; Article 3547; Gibbings, DJ., et al. (2007). BMC Immunol. 8; 12).

Physical form

Lyophilized from PBS containing D-Mannitol and Sucrose, normal appearance is a dried pellet. Reconstituted antibody solution is stable and functional as assessed by functional testing. Contains no biocide or preservatives, such as azide.

Reconstitution

1.0 mg/mL after reconstitution at 25 μL in PBS. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C. Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial. Reconstitute each vial by adding 25 μL of PBS. Protect from light.

Legal Information

ColorWheel is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

required but not provided

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service